Learn More
Description
- Superior durability with enhanced walls and crush-resistant turrets at the corners
- Easy handling with tabs to prevent stacked reservoirs from sticking together
Specifications
Specifications
| Capacity (English) | 0.17 oz. |
| Certifications/Compliance | DNase, RNase, and Pyrogen free quality certificate available upon request, Traceability Certificate available upon request |
| Color | White |
| Capacity (Metric) | 5 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| For Use With (Application) | Pipetting procedures |
| Packaging | Individually Wrapped |
| Quantity | 50 Pk. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
