Learn More
Research Products International Corp Reagent Reservoir, Sterile, Pyrogen Free,  25ml, 10 per Sleeve, 100 per Case
SDP
Catalog No. 50476544
Encompass
RPI Reagent Reservoirs have exclusive design innovations that improve accuracy and efficiency with multi-channel pipetting. Features include: four corner pouring spouts, trimmed base corners, molded graduations, wide base, sturdy construction and deep well micro-trough. These reservoirs are available in a non-sterile bulk pack for routine applications.
Ideal for PCR and other multi-channel pipetting applications
Deep Well micro-trough allows maximum sample recovers
Fit all 8 and 12 channel pipettors
Catalog No. 50-476-544 Supplier Research Products International Corp Supplier No. 248221
RPI Reagent Reservoirs have exclusive design innovations that improve accuracy and efficiency with multi-channel pipetting. Features include: four corner pouring spouts, trimmed base corners, molded graduations, wide base, sturdy construction and deep well micro-trough. These reservoirs are available in a non-sterile bulk pack for routine applications.
Ideal for PCR and other multi-channel pipetting applications
Deep Well micro-trough allows maximum sample recovers
Fit all 8 and 12 channel pipettors