Learn More
Research Products International Corp Reagent Reservoir Basins, 25ml Capacity, Sterile, Wrapped 5 per Pack, 100 per Case
SDP
Catalog No. 501367547
Encompass
Ideal reagent reservoir basins are designed with narrow 'V' bottom for complete sample aspiration. Reagent Basins accept 8 or 12 channel pipettes and feature rigid, durable walls with pour spouts to facilitate spill-free pouring. Unique packaging offers quick and easy open tear strip.
Catalog No. 50-136-7547 Supplier Research Products International Corp Supplier No. 246281
Ideal reagent reservoir basins are designed with narrow 'V' bottom for complete sample aspiration. Reagent Basins accept 8 or 12 channel pipettes and feature rigid, durable walls with pour spouts to facilitate spill-free pouring. Unique packaging offers quick and easy open tear strip.