Learn More
Research Products International Corp Reagent Reservoir, 25ml, Non-Sterile, Bulk 100/cs
SDP
Catalog No. 501978011
Encompass
RPI Reagent Reservoirs have exclusive design innovations that improve accuracy and efficiency with multi-channel pipetting. Features include: four corner pouring spouts, trimmed base corners, molded graduations, wide base, sturdy construction and deep well micro-trough. These reservoirs are available in a non-sterile bulk pack for routine applications.
Ideal for PCR and other multi-channel pipetting applications
Deep Well micro-trough allows maximum sample recovers
Fit all 8 and 12 channel pipettors
Non-Sterile
Catalog No. 50-197-8011 Supplier Research Products International Corp Supplier No. 248229
RPI Reagent Reservoirs have exclusive design innovations that improve accuracy and efficiency with multi-channel pipetting. Features include: four corner pouring spouts, trimmed base corners, molded graduations, wide base, sturdy construction and deep well micro-trough. These reservoirs are available in a non-sterile bulk pack for routine applications.
Ideal for PCR and other multi-channel pipetting applications
Deep Well micro-trough allows maximum sample recovers
Fit all 8 and 12 channel pipettors
Non-Sterile