Learn More
Research Products International Corp Reagent Reservoir Basins, 25ml Capacity, Sterile, Individually Wrapped, 50 per Case
SDP
Catalog No. 501367546
Encompass
Ideal reagent reservoir basins are designed with narrow 'V' bottom for complete sample aspiration. Reagent Basins accept 8 or 12 channel pipettes and feature rigid, durable walls with pour spouts to facilitate spill-free pouring. Unique packaging offers quick and easy open tear strip.
Catalog No. 50-136-7546 Supplier Research Products International Corp Supplier No. 246280
Add to Cart
Add to Cart
Ideal reagent reservoir basins are designed with narrow 'V' bottom for complete sample aspiration. Reagent Basins accept 8 or 12 channel pipettes and feature rigid, durable walls with pour spouts to facilitate spill-free pouring. Unique packaging offers quick and easy open tear strip.