Learn More
Stellar Scientific Reagent Reservoir, 25mL, Sterile, 5-pack, 200/cs
SDP
Catalog No. NC1761621
Encompass_Preferred
Corner "bump-outs" which make it easy to separate stacked reservoirs. RNase/DNase and human pyrogen free. A sharp "V" base that drives reagents towards the bottom of the trough for easy retrieval. Pour-off spouts on each corner for returning excess reagent and saving you money. Crisp graduation lines for visualizing volume.
Catalog No. NC1761621 Supplier Stellar Scientific Supplier No. R10255
Corner "bump-outs" which make it easy to separate stacked reservoirs. RNase/DNase and human pyrogen free. A sharp "V" base that drives reagents towards the bottom of the trough for easy retrieval. Pour-off spouts on each corner for returning excess reagent and saving you money. Crisp graduation lines for visualizing volume.