Learn More
CELLTREAT Scientific Products 50mL Reagent Reservoir, Polystyrene, Individually Wrapped, Sterile
SDP
Catalog No. 50201987
Encompass_Preferred
CELLTREAT Scientific 50mL Reagent Reservoir. Reagent Reservoirs are designed to assist in repetitive liquid transfer. White color enhances solution visibility. Angled design promotes funneling of liquid to maximize retrieval. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped/bag, 80/case.
Catalog No. 50-201-987 Supplier CELLTREAT Scientific Products Supplier No. 229290
CELLTREAT Scientific 50mL Reagent Reservoir. Reagent Reservoirs are designed to assist in repetitive liquid transfer. White color enhances solution visibility. Angled design promotes funneling of liquid to maximize retrieval. Rnase/Dnase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged sterile, individually wrapped/bag, 80/case.