Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Reagent Reservoir, Bulk wrap PROMO

Catalog No. 14127074PM
Change view
Click to view available options
Capacity (Metric):
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
14-127-074PM 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-127-074PM Supplier Fisherbrand™ Supplier No. 120690F2PM

Designed for precision

  • Superior durability with enhanced walls and crush-resistant turrets at the corners
  • Easy handling with tabs to prevent stacked reservoirs from sticking together

Specifications

Capacity (Metric) 50 mL
Dimensions (L x W x H) 4.5 x 1.1 x 1.4 in.
Material PVC
Sterility Non-sterile
Type Reagent Reservoir
Packaging Bulk
Quantity 100
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape V-bottom
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.