Learn More
Analytical Sales & Services Reagent Reservoir Lid
Catalog No. 5000806674
Encompass
Reagent Reservoir Lid\n• Medical Grade Polystyrene\n• Size: 127.8 x 85 x 9\n• Fits standard size footprint reservoirs\n• Color: clear\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 300 (10 per sleeve)
Catalog No. 50-008-06674 Supplier Analytical Sales & Services Supplier No. 69505
Reagent Reservoir Lid\n• Medical Grade Polystyrene\n• Size: 127.8 x 85 x 9\n• Fits standard size footprint reservoirs\n• Color: clear\n• Certified DNase/RNase and Pyrogen Free\n\nQty/Pkg: 300 (10 per sleeve)