Learn More
Research Products International Corp Reagent Reservoir without Lid, Non-Sterile, 175 ml, 10 per Case
SDP
Catalog No. 50212184
Encompass
Reagent reservoirs are reusable. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 ml.
Catalog No. 50-212-184 Supplier Research Products International Corp Supplier No. 140196
Reagent reservoirs are reusable. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 ml.