Learn More
Sigma Aldrich Fine Chemicals Biosciences Reagent Reservoir reservoi
Catalog No. 502901723
Encompass
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.
Catalog No. 50-290-1723 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. HS120641100EA
Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures. Graduation markingson the top edge and inner walls allows measurements of liquid to be easily identified and reduces waste of expensive reagents. Pour-off spouts design on all four corners enables easy spill-free pouring and reduces chance of dripping.