Learn More
VistaLab Reagent Reservoir, Small Footprint, Bulk, 50 mL
SDP
Catalog No. 09035114 Shop All VistaLab Technologies Inc. Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
09-035-114 Non-sterile
09-035-111 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 09-035-114 Supplier VistaLab Supplier No. 40541006SF
Add to Cart
Add to Cart
VistaLab Reservoir small footprint design offers a narrower base and deeper basin to save bench space and is uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.
  • VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.
  • Trough-within-a-trough design to recover the 'last few drops'.
  • Pour-off spouts at every corner.
  • Narrower base and deeper basin to save bench space.
  • Free of animal products.
  • Sterile reservoirs are certified RNase/DNase free and non-pyrogenic.
  • Well-marked graduated increments on the inside wall.
  • Bulk or individually wrapped packaging.
  • Molded corner structures allow bulk packaged units to be easily separated for use.
  • Clearly marked for recycling.
  • Available in variety of sizes.
  • 12-channel polypropylene reservoir with lid is autoclavable.
Specifications
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Non-sterile
Type Reagent Reservoir
Autoclavable Not Autoclavable
Channels 1
Disposable Reusable
Bottom Shape Conical