Learn More
VistaLab Reagent Reservoir, Small Footprint, Individually Wrapped, Sterile, 50 mL
SDP
Catalog No. 09035112 Shop All VistaLab Technologies Inc. Products
Catalog No. 09-035-112 Supplier VistaLab Supplier No. 30541007SF
Only null left
Add to Cart
Add to Cart
VistaLab Reservoir small footprint design offers a narrower base and deeper basin to save bench space and is uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.
  • VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.
  • Trough-within-a-trough design to recover the 'last few drops'.
  • Pour-off spouts at every corner.
  • Narrower base and deeper basin to save bench space.
  • Free of animal products.
  • Sterile reservoirs are certified RNase/DNase free and non-pyrogenic.
  • Well-marked graduated increments on the inside wall.
  • Bulk or individually wrapped packaging.
  • Molded corner structures allow bulk packaged units to be easily separated for use.
  • Clearly marked for recycling.
  • Available in variety of sizes.
  • 12-channel polypropylene reservoir with lid is autoclavable.
Specifications
Capacity (Metric) 50 mL
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Quantity 50/Cs.
Autoclavable Not Autoclavable
Channels 1
Disposable Reusable
Bottom Shape Conical