Learn More
VistaLab Reagent Reservoir, Small Footprint, Individually Wrapped, Sterile, 50 mL

Shop All VistaLab Technologies Inc. Products


Description
- VistaLab reservoirs are uniquely treated to provide the lowest residual volume in the industry, meaning maximal reagent recovery, up to 67% better than comparable reservoirs.
- Trough-within-a-trough design to recover the 'last few drops'.
- Pour-off spouts at every corner.
- Narrower base and deeper basin to save bench space.
- Free of animal products.
- Sterile reservoirs are certified RNase/DNase free and non-pyrogenic.
- Well-marked graduated increments on the inside wall.
- Bulk or individually wrapped packaging.
- Molded corner structures allow bulk packaged units to be easily separated for use.
- Clearly marked for recycling.
- Available in variety of sizes.
- 12-channel polypropylene reservoir with lid is autoclavable.
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Material | Polystyrene |
| Sterility | Sterile |
| Type | Reagent Reservoir |
| Quantity | 100/Cs. |
| Autoclavable | Not Autoclavable |
| Channels | 1 |
| Disposable | Reusable |
| Bottom Shape | Conical |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.