Learn More
Hudson Control Group, Inc. REAGENT RESERVOIR W/3 BAFFLES
SDP
Catalog No. NC9585738
Encompass
Reagent Reservoir (Quadra 96 accessory), Standard Height (1.90 in. ), 3 baffles with dimples, 300mL, Molded of inert polypropylene; working volume of 300mL, Used on 96-well Quadras.
Catalog No. NC9585738 Supplier Hudson Control Group, Inc. Supplier No. 19613000
Reagent Reservoir (Quadra 96 accessory), Standard Height (1.90 in. ), 3 baffles with dimples, 300mL, Molded of inert polypropylene; working volume of 300mL, Used on 96-well Quadras.