Learn More
Research Products International Corp Reagent Reservoir with Lid, Non-Sterile, 175 ml Capacity, 10 per Case
SDP
Catalog No. 50212183
Encompass
Reagent reservoirs are reusable. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight and twelve place multiple pipettors with minimal reagent waste. Lids are removable, attached with ball and socket hinges, and latch tightly for secure storage of left-over reagents. With lids closed, reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 ml.
Catalog No. 50-212-183 Supplier Research Products International Corp Supplier No. 140195
Reagent reservoirs are reusable. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight and twelve place multiple pipettors with minimal reagent waste. Lids are removable, attached with ball and socket hinges, and latch tightly for secure storage of left-over reagents. With lids closed, reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear, chemical-resistant, autoclavable polypropylene. Molded graduation marks provide volume indication at 25, 50, 75, 100, 125, and 150 ml.