Learn More
Bellco Glass, Inc. Reagent Reservoir with Caps, 60mL, Sterile, 5/pk, 25/cs
SDP
Catalog No. 502330860
Encompass_Preferred
Reagent Reservoir with Caps, 60mL, Sterile, 5/pk, 25/cs. For liquid storage in experiments, it is convenient for repeated addition of liquid. Can be used with single or multi-channel pipettes. • V-shape Bottom, used for cell culture and immunoassay experiments• For single and multi-channel pipettes• USP VI PP material• Reusable, autoclave at 121℃• Special designed lid to prevent contamination• Clear graduations• DNase/RNase free, Non-pyrogenic
Catalog No. 50-233-0860 Supplier Bellco Glass, Inc. Supplier No. 451400413
Reagent Reservoir with Caps, 60mL, Sterile, 5/pk, 25/cs. For liquid storage in experiments, it is convenient for repeated addition of liquid. Can be used with single or multi-channel pipettes. • V-shape Bottom, used for cell culture and immunoassay experiments• For single and multi-channel pipettes• USP VI PP material• Reusable, autoclave at 121℃• Special designed lid to prevent contamination• Clear graduations• DNase/RNase free, Non-pyrogenic