Learn More
Sigma Aldrich Fine Chemicals Biosciences Reagent Reservoir with Hin
Catalog No. 502901272
Encompass
Volume graduations to 150 mLA reinforced base that wont wobble on the counter preventing spillsA new easy-to-open latch that can be manipulated with one handGreater clarity for improved visibility of contentsReagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents.With lids closed reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear chemical-resistant autoclavable polypropylene. Molded graduation marks provide volume indication at 25 50 75 100 125 and 150 mL
Catalog No. 50-290-1272 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z74407410EA
Volume graduations to 150 mLA reinforced base that wont wobble on the counter preventing spillsA new easy-to-open latch that can be manipulated with one handGreater clarity for improved visibility of contentsReagent reservoirs are reusable -- an environmentally sound alternative to disposable reagent reservoirs. Their extra-large capacity means fewer refills and time saved. The V-shaped basin is compatible with both eight- and twelve-place multiple pipettors with minimal reagent waste. Lids are removable -- attached with ball and socket hinges -- and latch tightly for secure storage of left-over reagents.With lids closed reagent reservoirs are stackable. A perimeter ridge on the lid prevents sliding. Construction is clear chemical-resistant autoclavable polypropylene. Molded graduation marks provide volume indication at 25 50 75 100 125 and 150 mL