Learn More
Chemglass Life Sciences Reagent Reservoirs, 10mL, Bulk Packed, Sterile
SDP
Catalog No. 501532336
Encompass
Reagent Reservoirs, 10mL, Bulk Packed, Non-Sterile. Disposable 10mL reagent reservoirs are made from sturdy white polystyrene. The compact size minimizes waste when working with smaller volumes due to reduced width/length which provides less surface area for hold up. Reservoirs have been designed with trimmed base corners, deeper pour-off spouts for spill-free emptying, and a deeper, narrower ¿trough within a trough¿ to maximize reagent recovery. Reservoirs are available Sterile and Non-Sterile.
Catalog No. 50-153-2336 Supplier Chemglass Life Sciences Supplier No. CLS3797B010
Reagent Reservoirs, 10mL, Bulk Packed, Non-Sterile. Disposable 10mL reagent reservoirs are made from sturdy white polystyrene. The compact size minimizes waste when working with smaller volumes due to reduced width/length which provides less surface area for hold up. Reservoirs have been designed with trimmed base corners, deeper pour-off spouts for spill-free emptying, and a deeper, narrower ¿trough within a trough¿ to maximize reagent recovery. Reservoirs are available Sterile and Non-Sterile.