Learn More
Chemglass Life Sciences Reagent Reservoirs, 10mL, Individually Wrapped, Sterile
SDP
Catalog No. 501532337
Encompass
Reagent Reservoirs, 10mL, Individually Wrapped, Sterile. Disposable 10mL reagent reservoirs are made from sturdy white polystyrene. The compact size minimizes waste when working with smaller volumes due to reduced width/length which provides less surface area for hold up. Reservoirs have been designed with trimmed base corners, deeper pour-off spouts for spill-free emptying, and a deeper, narrower trough within a trough to maximize reagent recovery. Material: Polystyrene. Minimize Solvent Waste- Compact Trough Design. RNase/DNase Certified. Pyrogen Free. Animal Free. Marked for Recycling.
Catalog No. 50-153-2337 Supplier Chemglass Life Sciences Supplier No. CLS3797B011
Reagent Reservoirs, 10mL, Individually Wrapped, Sterile. Disposable 10mL reagent reservoirs are made from sturdy white polystyrene. The compact size minimizes waste when working with smaller volumes due to reduced width/length which provides less surface area for hold up. Reservoirs have been designed with trimmed base corners, deeper pour-off spouts for spill-free emptying, and a deeper, narrower trough within a trough to maximize reagent recovery. Material: Polystyrene. Minimize Solvent Waste- Compact Trough Design. RNase/DNase Certified. Pyrogen Free. Animal Free. Marked for Recycling.