Learn More
Andwin Scientific Disposable Pipetting Reservoirs, 50 mL, Sterile, Individually Wrapped, 100/cs
SDP
Catalog No. NC0588685
Encompass_Preferred
Disposable Pipetting Reservoirs are designed for use in multichannel pipetting applications, and will accommodate all types of pipettors up to 12-channel width. Pipetting reservoirs feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.
Catalog No. NC0588685 Supplier Andwin Scientific Supplier No. 89094686
Add to Cart
Add to Cart
Disposable Pipetting Reservoirs are designed for use in multichannel pipetting applications, and will accommodate all types of pipettors up to 12-channel width. Pipetting reservoirs feature a trough design, graduation marks on interior walls, and pour-off spouts on each corner. All reservoirs are lot-certified, nonpyrogenic, and DNase- and RNase-free.