Learn More
Hach Company Silica Reagent Set, HR, 25 mL
Supplier:  Hach Company 2244300
Encompass
For high range silica determination by the Silicomolybdate method (Hach method 8185). Range: up to 100 mg/L SiO2. Sample Size: 25 mL. 100 tests
Catalog No. 50-199-5627