Learn More
Hach Company Alkalinity Reagent Set, Digital Titrator
Supplier:  Hach Company 2271900
Encompass
For alkalinity determination by sulfuric acid titration with the Digital Titrator. Hach method 8203. Range: 10-4,000 mg/L CaCO3 Set contains Phenolphthalein Indicator Powder Pillows, Bromcresol Green-Methyl Red Indicator Powder Pillows and Sulfuric Acid Digital Titrator Cartridges, 0.1600 N and 1.600 N. Approximately 40-100 tests.
Catalog No. 50-199-5690