Learn More
Hach Company Hardness Reagent Set, Calmagite Colorimetric
Supplier:  Hach Company 2319900
Encompass
For determination of Calcium and Magnesium Hardness by the Calmagite Colorimetric method. Hach Method 8030. Range: 0.05 - 4.00 mg/L as CaCO3. Reagent set contains: Alkali Solution, Calcium/Magnesium Indicator Solution, EDTA Solution and EGTA Solution for 100 tests.
Catalog No. 50-199-5795