Learn More
Hach Company Hardness (Calcium and Total) Reagent Set, Digital Titrator
Catalog No. 501995692 Shop All Hach Company Products
Encompass
For Calcium and Total Hardness determination by EDTA Digital Titrator titration. Ranges: 10-160 and 100-4000 mg/L as CaCO3. Reagent Set Contains: Buffer Solution (Hardness 1), CalVer 2 Calcium Indicator Powder Pillows, ManVer 2 Hardness Indicator Powder Pillows, Potassium Hydroxide Solution (8 N), EDTA Titration Cartridges (1 each) (0.800 and 0.0800 M).
Catalog No. 50-199-5692 Supplier Hach Company Supplier No. 2272100
For Calcium and Total Hardness determination by EDTA Digital Titrator titration. Ranges: 10-160 and 100-4000 mg/L as CaCO3. Reagent Set Contains: Buffer Solution (Hardness 1), CalVer 2 Calcium Indicator Powder Pillows, ManVer 2 Hardness Indicator Powder Pillows, Potassium Hydroxide Solution (8 N), EDTA Titration Cartridges (1 each) (0.800 and 0.0800 M).