Learn More
Hach Company Carbon Dioxide Reagent Set, Digital Titrator
Supplier:  Hach Company 2272700
Encompass
For determination of Carbon Dioxide by Digital Titrator titration. Hach Method 8205. Range: 10 - 1000 mgL as CO2 Set includes: Phenolphthalein Pillows and two Sodium Hydroxide Digital Titrator Cartridges (0.3636 N and 3.3636 N). Approximately 40-100 tests
Catalog No. 50-199-5698