Learn More
Hach Company Carbon Dioxide Reagent Set for Model CA-23 Test Kit
Supplier:  Hach Company 2437500
Encompass
For determination of Carbon Dioxide by drop-count titration. Replacement reagent set for Hach Test Kit Model CA-23. Includes: Phenolphthalein Indicator Solution and Sodium Hydroxide Standard Solution (0.01 N).
Catalog No. 50-199-5991