Learn More
Hach Company Chloride Reagent Set, LR, Digital Titrator
Supplier:  Hach Company 2444100
Encompass
For determination of Chloride by argentometric Digital Titrator titration. Range: 10 - 100 mg/L Cl-. Reagent Set contains Chloride 2 Indicator Powder Pillows and Silver Nitrate Digital Titrator Cartridges (2) 0.2256 N. Approximately 100 tests
Catalog No. 50-199-6006