Learn More
Hach Company Chlorine Dioxide Reagent Set, Chlorophenol Red
Supplier:  Hach Company 2242300
Encompass
For determination of Chlorine Doxide by the Chlorophenol Red Method. Hach method 8065. Range: 0.01 - 1.00 mg/L ClO2. Set contains Chlorine Dioxide Reagent 1, 2, and 3, and Dechlorinating Reagent Powder Pillows for 100 tests. Sample Size: 50 mL
Catalog No. 50-199-5610