Learn More
Hach Company Chlorine (Free and Total) Reagent Set, DPD 5 mL
Catalog No. 501995997 Shop All Hach Company Products
Encompass
For determination of Free and Total Chlorine by the DPD method, using visual test kits. Set contains: DPD Free Chlorine PP, pk/100 and DPD Total Chlorine PP pk/100. Range: 0 - 3.5 mg/L as Cl₂. Sample Size: 5 mL.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-5997 Supplier Hach Company Supplier No. 2438800
For determination of Free and Total Chlorine by the DPD method, using visual test kits. Set contains: DPD Free Chlorine PP, pk/100 and DPD Total Chlorine PP pk/100. Range: 0 - 3.5 mg/L as Cl₂. Sample Size: 5 mL.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.