Learn More
Hach Company Chlorine Reagent Set, DPD Free and Total Chlorine, 25 mL sample
Catalog No. 501996004 Shop All Hach Company Products
Encompass
Reagent Set, Chlorine (Free & Total DPD Colorimetric Method). 100 tests each, for 25 mL samples.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. 50-199-6004 Supplier Hach Company Supplier No. 2440400
Reagent Set, Chlorine (Free & Total DPD Colorimetric Method). 100 tests each, for 25 mL samples.Boric Acid free!*This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.