Learn More
Hach Company CL17 Total Chlorine Reagent Set
Supplier:  Hach Company 2557000
Encompass
Total Chlorine Reagent Set (includes DPD Indicator, and Total Chlorine Indicator and Buffer Solutions). Used with the CL17 Analyzer and bulk analytical methods. This product has not been evaluated to test for chlorine and chloramines in medical applications in the United States.
Catalog No. NC9405770