Learn More
Hach Company Chlorine (Total) Reagent Set for CN-65 Test Kit
Supplier:  Hach Company 2438600
Encompass
For determination of Total Chlorine by Drop Count Titration. Replacement reagent set for CN-65 Test Kit (225401). Set includes: Sulfamic Acid Powder Pillows, Sulfite 1 Reagent Powder Pillows and Sodium Thiosulfate Standard Solution for approximately 100 tests.
Catalog No. 50-199-5996