Learn More
Hach Company Chlorine (Total) Reagent Set for CN-21P
Supplier:  Hach Company 2437700
Encompass
For determination of Chlorine (Total) by thiosulfate drop-count titration. Replacement reagent set for CN-21P Test Kit. Set includes Sulfamic Acid Powder Pillows, Potassium Iodide Powder Pillows and Sodium Thiosulfate Standard Solution (0.0246 N). Approx. 100 tests.
Catalog No. 50-199-5993