Learn More
Hach Company Cobalt / Nickel Reagent Set, PAN Method, 25-mL sample
Supplier:  Hach Company 2242600
Encompass
For determination of Cobalt and Nickel by the PAN method. Hach Method 8078. Sample Size: 25 mL Range: 0.01 - 2.00 mg/L Co, 0.006 - 1.000 mg/L Ni. Reagent Set includes: EDTA Reagent Powder Pillows, Phthalate-Phosphate Reagent Powder Pillows and PAN Indicator Solution (0.3%) for 100 tests.
Catalog No. 50-199-5613