Learn More
Hach Company Copper Reagent Set, Porphyrin, 10 mL
Supplier:  Hach Company 2603300
Encompass
For Copper determination by the Porphyrin Method. Hach Method 8143 Range: 2 - 210 μg/L Cu. Sample Size: 10 mL Reagent set Includes Copper Masking Reagent Powder Pillows, Porphyrin 1 Reagent Powder Pillows and Porphyrin 2 Reagent Powder Pillows for approximately 100 tests.
Catalog No. 50-199-6314