Learn More
Hach Company Copper Reagent Set, Porphyrin, 25 mL
Supplier:  Hach Company 2242700
Encompass
For determination of copper by the Porphyrin Method. Hach Method 8143. Range: 2 - 210 μg/L Sample Size: 25 mL. Reagent Set includes: Copper Masking Reagent Powder Pillows, Porphyrin 1 and 2 Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-5614