Learn More
Hach Company Cyanide Reagent Set, CyaniVer, 10 mL
Supplier:  Hach Company 2430200
Encompass
For determination of Cyanide by the Pyridine-Pyrazalone Method. Hach Method 8027. Range: 0.002 - 0.240 mg/L CN-. Sample Size: 10 mL. Reagent Set Includes: CyaniVer 3, 4, and 5 Cyanide Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-5974
 Restricted Item