Learn More
Hach Company Dissolved Oxygen Reagent Set, 60 mL sample
Supplier:  Hach Company 2439802
Encompass
For determination of Dissolved Oxygen by Drop Count Titration. Replacement reagent set for OX-2P Test Kit (1469-00). Contains: Dissolved Oxygen 1, 2, and 3 Reagent Powder Pillows and Sodium Thiosulfate Standard (0.0109 N) for about 100 tests.
Catalog No. 50-199-6000