Learn More
Hach Company FerroMo™ Iron Reagent Set
Supplier:  Hach Company 2544800
Encompass
For determination of iron in cooling waters containing molybdate-based treatments. Hach Method 8365. Range: 0.01 - 1.80 mg/L Fe Reagent Set Includes: FerroMo™ Reagent 1 Powder Pillows and FerroMo™ Reagent 2 Powder Pillows for 100 tests.
Catalog No. 50-199-6201