Learn More
HANNA INSTRUMENTS INC Reagent Set for HI83730 photometer
SDP
Catalog No. NC1809401
Encompass
Reagent set for the HI83730 photometer for the determination of peroxide value in olive oils. A user can accurately determine the peroxide value of olive oil within a 0.0 to 25.0 meq O2/kg range using the HI83730-20 ready made reagents.Ready madeMarked with lot number for traceabiltyFor use with the HI83730 photometer for the determination of peroxide value in olive oils
Catalog No. NC1809401 Supplier HANNA INSTRUMENTS INC Supplier No. HI8373020
Reagent set for the HI83730 photometer for the determination of peroxide value in olive oils. A user can accurately determine the peroxide value of olive oil within a 0.0 to 25.0 meq O2/kg range using the HI83730-20 ready made reagents.Ready madeMarked with lot number for traceabiltyFor use with the HI83730 photometer for the determination of peroxide value in olive oils