Learn More
Hach Company Hardness (Total) Reagent Set for HA-71A Test Kit
Supplier:  Hach Company 2437800
Encompass
For determination of Total Hardness by the Drop Count EDTA Titration Method. Replacement reagent set for HA-71A Test Kit. Set contains Buffer Solution (Hardness 1), Hardness 2 and Hardness 3 Test Solutions for approximately 100 tests.
Catalog No. 50-199-5994