Learn More
Hach Company Hardness (Total) Reagent Set for 5-EP Test Kit
Supplier:  Hach Company 2437900
Encompass
For determination of Total Hardness by the Drop Count EDTA Titration Method. Replacement reagent set for 5-EP Test Kit. Set contains UniVer 3 Powder Pillows and Hardness 3 Test Solution for approximately 100 tests.
Catalog No. 50-199-5995