Learn More
Hach Company Hardness (Calcium) Reagent Set, Buret
Supplier:  Hach Company 2447000
Encompass
Reagent set for the determination of Calcium Hardness by EDTA Buret Titration Method (Hach method 8222). EPA Accepted method. Range: 0-5,000 mg/L as CaCO3. Set contains CalVer 2 Calcium Indicator Powder Pillows, Potassium Hydroxide Solution (8 N) and TitraVer (EDTA) Titrant for up to approximately 100 tests.
Catalog No. 50-199-6019