Learn More
Hach Company Hardness (Total) Reagent Set, HR, Digital Titrator
Supplier:  Hach Company 2448100
Encompass
Reagent set for the determination of Total Hardness by EDTA Digital Titrator titration (Hach method 8213). Range: 100-4,000 mg/L as CaCO3. Set contains: Buffer Solution (Hardness 1), ManVer 2 Hardness Indicator Powder Pillows and 0.800 M EDTA Digital Titrator Cartridge for 40 - 100 tests.
Catalog No. 50-199-6025