Learn More
Hach Company Hardness (Calcium) Reagent Set, HR, Digital Titrator
Supplier:  Hach Company 2447500
Encompass
Reagent set for the determination of Calcium Hardness by EDTA Digital Titrator Titration Method (Hach method 8204). Range: 100-4000 mg/L as CaCO3. Set contains CalVer 2 Calcium Indicator Powder Pillows, Potassium Hydroxide Solution (8 N) and 0.800 M (EDTA) Digital Titrator Cartridge for approximately 40-100 tests.
Catalog No. 50-199-6021