Learn More
Hach Company Hardness (Calcium & Total Sequential) LR Reagent Set, Digital Titrator
Catalog No. 501996027 Shop All Hach Company Products
Encompass
For sequential determination of Calcium and Total Hardness by EDTA Digital Titrator titration. Hach method 8329. Range: 10-160 mg/L CaCO3. Set contains Buffer Solution (Hardness 1), CalVer2 Calcium Indicator Powder Pillows, ManVer2 Hardness Indicator Powder Pillows, Potassium Hydroxide Solution (8 N), Sulfuric Acid (5.25 N) and 0.0800 M EDTA Digital Titrator Cartridge. Approximately 40-100 tests.
Catalog No. 50-199-6027 Supplier Hach Company Supplier No. 2448600
For sequential determination of Calcium and Total Hardness by EDTA Digital Titrator titration. Hach method 8329. Range: 10-160 mg/L CaCO3. Set contains Buffer Solution (Hardness 1), CalVer2 Calcium Indicator Powder Pillows, ManVer2 Hardness Indicator Powder Pillows, Potassium Hydroxide Solution (8 N), Sulfuric Acid (5.25 N) and 0.0800 M EDTA Digital Titrator Cartridge. Approximately 40-100 tests.