Learn More
Hach Company Hydrogen Peroxide Reagent Set
Catalog No. 501996029 Shop All Hach Company Products
Encompass
For determination of Hydrogen Peroxide by drop count titration. Replacement reagent set for HYP-1 Test Kit (2291700). Approximately 100 Tests.
Catalog No. 50-199-6029 Supplier Hach Company Supplier No. 2449100
For determination of Hydrogen Peroxide by drop count titration. Replacement reagent set for HYP-1 Test Kit (2291700). Approximately 100 Tests.