Learn More
Hach Company LeadTrak™ Reagent Set, Fast Column Extraction Method, 20 tests
Catalog No. 501995891 Shop All Hach Company Products
Encompass
Reagent Set for Lead determination by the LeadTrak™ Fast Column Extraction. Hach Method 8317. Range: 5 to 150 µg/L as Pb. Sample Size: 100 mL. 20 tests/Reagent Set.
Catalog No. 50-199-5891 Supplier Hach Company Supplier No. 2375000
Reagent Set for Lead determination by the LeadTrak™ Fast Column Extraction. Hach Method 8317. Range: 5 to 150 µg/L as Pb. Sample Size: 100 mL. 20 tests/Reagent Set.