Learn More
Hach Company Manganese Reagent Set, HR, 10 mL
Supplier:  Hach Company 2430000
Encompass
For determination of Manganese by the Periodate Oxidation Method. USEPA accepted for reporting wastewater analysis using Hach Method 8034. Range: 0.1 - 20.0 mg/L Mn. Sample Size: 10 mL. Reagent Set includes Citrate Buffer Powder Pillows and Sodium Periodate Powder Pillows for 100 tests. To confirm accurate results, see appropriate standard solutions on the Consumables tab.
Catalog No. 50-199-5973